missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Dropping Pipettes for Soda-Lime Reagent Bottles

Manufactured from clear soda-lime glass

Brand:  MBL SBD100

482.60 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 15768057

  • 508.00 DKK / Pack of 5
Only null left
Estimated Shipment: 26-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from clear soda-lime glass

  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

14/23 Joint
Dropping pipettes with silicone teat
100 mL Bottle
Soda Lime Glass
Pasteur Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MBL™ Dropping Pipettes for Soda-Lime Reagent Bottles > With: Joint size 14/23; For use with (equipment): 100mL bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.