Få mere at vide
Mavoglurant racemate, MedChemExpress
Artikelnummer. 16430236
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
Pakningsstørrelse:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16430236 2 mg -
16408997 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16430236 Leverandør Medchem Express Leverandørnr. HY15257A
MedChemExpress Mavoglurant racemate (AFQ-056 racemate) is the racemate of Mavoglurant. Mavoglurant is a novel, non-competitive mGlu5 receptor antagonist.
Mavoglurant racemate (AFQ-056 racemate) is the racemate of Mavoglurant. Mavoglurant is a novel, non-competitive mGlu5 receptor antagonist.
Tekniske data
Kemisk navn eller materiale Mavoglurant racemate
CAS 1636881-61-2
Farve White
Renhedsgrad noter Research
Molekylær formel C19H23NO3
Mængde 2 mg
Synonym AFQ-056 racemate
Opløselighedsinformation DMSO : 50 mg/mL (159.55 mM; Need ultrasonic)
SMIL O=C(N1CC[C@@]2([H])[C@@](C#CC3=CC=CC(C)=C3)(O)CCC[C@@]12[H])OC
Molekylvægt (g/mol) 313.39
Formel vægt 313.39
Procent renhed 98.44%
Grad Research
Fysisk form Solid