Learn More
Masterflex™ Silicone Peroxide Softwall Standard Tubing
Softwall silicone peroxide tubing for biological applications. Masterflex™ Silicone Peroxide Softwall Standard Tubing has excellent biological compatibility.
448.00 DKK - 1315.00 DKK
Specifications
Product Type | Standard Tubing |
---|---|
Certifications/Compliance | US Pharmacopoea XXIII Cl.Vl FDA 21 CFR, Part 177.2600 |
Autoclavable | Autoclavable |
Material | Silicone (Peroxide-cured) |
Overall Length (Metric) | 7.5 m |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
10433232
|
Masterflex™
MF0044ST |
0.8mm I.D., 4.0mm O.D. |
795.00 DKK
7.50m |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
10303512
|
Masterflex™
MF0037ST |
3.2mm I.D., 6.4mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10654213
|
Masterflex™
MF0045ST |
4.8mm I.D., 8.0mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10614023
|
Masterflex™
MF0046ST |
6.4mm I.D., 9.6mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10674403
|
Masterflex™
MF0047ST |
8.0mm I.D., 11.2mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10154742
|
Masterflex™
MF0035ST |
1.6mm I.D., 4.8mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- For biological applications.
- Autoclavable.
- Low gas permeability.
- Excellent biological compatibility.
- Great physical compression capability.
- Not prone to mold.
- Non-toxic.
- Waterproof and resistant to ozone, radiation and sunlight.
- Resistant to fungus.
- Odorless.
- Not recommended for concentrated solvents, oils, acids or diluted sodium hydroxide.
- Relatively high gas permeability.
- Applications
- Acids: Limited
- Alkaline solutions: Not recommended
- Solvents: Not recommended
- Pressure: Not recommended
- Vacuum: Good.
- Viscous media: Good
- Sterile media: Fair
- Complies with: US Pharmacopoea XXIII Cl.Vl FDA 21 CFR175.300
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15psi) and 121°C.
- Radiation: Irradiate at up to 2.5 mrad.
- Gas: Not recommended to sterilize with ethylene oxide.
Specifications
Standard Tubing | |
Autoclavable | |
7.5 m | |
Biological applications | |
CO2 25147, O2 4715, N2 2284 | |
Clear | |
238°C |
US Pharmacopoea XXIII Cl.Vl FDA 21 CFR, Part 177.2600 | |
Silicone (Peroxide-cured) | |
1.6 mm | |
-51°C | |
Not recommended for concentrated solvents, oils, acids or diluted sodium hydroxide. Relatively high gas permeability. | |
Good |