missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Marine Label Tape for BradyJet J4000 and J5000, BS 5609
Don’t let a label on the final product sink your efficiency. With BS 5609 Marine Labels and the BradyJet J4000 or J5000 Printer, your labels stay stuck, even at sea.
1515.00 DKK - 2805.00 DKK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Acrylic |
Color | White |
Finish | Matte |
Material | Inkjet Receptive Polyester |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
17469202
|
Brady™
150760 |
57.2 mm |
1515.00 DKK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17479202
|
Brady™
150761 |
101.6 mm |
2090.00 DKK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17489202
|
Brady™
150762 |
152.4 mm |
2515.00 DKK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17499202
|
Brady™
150763 |
203.2 mm |
2805.00 DKK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- Keep it colorful- this material runs through the BradyJet J4000 or J5000 Inkjet printer for high-resolution, full-color printing
- An ink cartridge is required to print this material
Specifications
Not Autoclavable | |
White | |
Inkjet Receptive Polyester | |
Non-sterile | |
BradyJet J4000 and J5000 Color Label Printers | |
1 Tape | |
0.22 mm |
Acrylic | |
Matte | |
Inkjet | |
General Identification, Hazard Communication Identification, Marine Labeling | |
1 | |
-40°C to +60°C | |
30.5 mm |