Learn More
Brady™ Marine Label Tape for BradyJet J4000 and J5000, BS 5609
Don’t let a label on the final product sink your efficiency. With BS 5609 Marine Labels and the BradyJet J4000 or J5000 Printer, your labels stay stuck, even at sea.
Brand: Brady™ 150760
Additional Details : Weight : 0.46170kg
Description
- Keep it colorful- this material runs through the BradyJet J4000 or J5000 Inkjet printer for high-resolution, full-color printing
- An ink cartridge is required to print this material
Specifications
Not Autoclavable | |
White | |
Inkjet Receptive Polyester | |
Non-sterile | |
BradyJet J4000 and J5000 Color Label Printers | |
1 Tape | |
0.22 mm | |
57.2 mm |
Acrylic | |
Matte | |
Inkjet | |
General Identification, Hazard Communication Identification, Marine Labeling | |
1 | |
-40°C to +60°C | |
30.5 mm |