Få mere at vide
Manidipine dihydrochloride, Thermo Scientific™
Calcium channel blocker
Brand: Thermo Scientific Alfa Aesar J60496.MB
| Mængde | 25mg |
|---|
Se flere versioner af dette produkt
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 89226-75-5 | |
| 683.627 | |
| JINNGBXKBDUGQT-UHFFFAOYSA-N | |
| 150762 | |
| CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl |
| C35H40Cl2N4O6 | |
| MFCD00896434 | |
| manidipine hydrochloride | |
| 5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride |
Tekniske data
| Manidipine dihydrochloride | |
| C35H40Cl2N4O6 | |
| 25mg | |
| 14,5743 | |
| Soluble in DMSO; Slightly soluble in water and ethanol /n | |
| CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl | |
| 683.627 | |
| 683.63 |
| 89226-75-5 | |
| MFCD00896434 | |
| UN2811 | |
| manidipine hydrochloride | |
| JINNGBXKBDUGQT-UHFFFAOYSA-N | |
| 5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride | |
| 150762 |