missing translation for 'onlineSavingsMsg'
Få mere at vide

Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific™

1067.18 DKK - 2649.29 DKK

Kemiske identifikatorer

CAS 7790-33-2
MDL nummer MFCD00016225
Se flere specifikationer

Produkter 2
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15497425
Dokumentvisning
Thermo Scientific Alfa Aesar
047166.03
1g
1067.18 DKK
1g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15407435
Dokumentvisning
Thermo Scientific Alfa Aesar
047166.06
5g
2649.29 DKK
5g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
Tekniske data

Kemiske identifikatorer

7790-33-2
MFCD00016225

Tekniske data

Manganese(II) iodide
7790-33-2
MFCD00016225
Soluble in water.
99.99%
Ampule
5.0 g/mL
Ultra dry
(metals basis)
14,5732
308.75
Odorless
Air, light, and moisture sensitive
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 232-201-6

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.