Få mere at vide
(±)-Mandelic-2,3,4,5,6-d5 Acid, TRC
Artikelnummer. 17907679
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17907679 1 g
17969109 250 mg
17907499 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17907679 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1625271G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70838-71-0
Molekylær formel C8 2H5 H3 O3
Molekylvægt (g/mol) 157.18
Synonym Benzene-d5-acetic acid, α-hydroxy-, α-Hydroxybenzene-d5-acetic acid, (±)-Mandelic-2,3,4,5,6-d5 acid, Mandelic acid-d5, Mandelic acid D5 (phenyl-D5), 2-Hydroxy-2-(phenyl-d5)acetic acid, 2-Hydroxy-2-pentadeuteriophenylacetic acid
IUPAC navn 2-hydroxy-2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid
SMIL [2H]c1c([2H])c([2H])c(C(O)C(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale (±)-Mandelic-2,3,4,5,6-d5 Acid
Mængde 1 g
InChI formel InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/i1D,2D,3D,4D,5D
Formel vægt 157.0787