CAS RN 70838-71-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(±)-Mandelic-2,3,4,5,6-d5 Acid, TRC
CAS: 70838-71-0 Molekylær formel: C8 2H5 H3 O3 Molekylvægt (g/mol): 157.18 Synonym: Benzene-d5-acetic acid, α-hydroxy-,α-Hydroxybenzene-d5-acetic acid,(±)-Mandelic-2,3,4,5,6-d5 acid,Mandelic acid-d5,Mandelic acid D5 (phenyl-D5),2-Hydroxy-2-(phenyl-d5)acetic acid,2-Hydroxy-2-pentadeuteriophenylacetic acid IUPAC navn: 2-hydroxy-2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid SMIL: [2H]c1c([2H])c([2H])c(C(O)C(=O)O)c([2H])c1[2H]
2
(±)-Mandelic-2,3,4,5,6-d5 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.