Få mere at vide
Maltol-d3, TRC
Artikelnummer. 17938969
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17938969 2.5 mg 2.5mg
17908669 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17938969 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1601322.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132331-92-1
Molekylær formel C6H3D3O3
Molekylvægt (g/mol) 129.13
Synonym 3-Hydroxy-2-methyl-4H-pyran-4-one, 2-Methyl-3-hydroxy-4-pyrone-d3, 2-Methyl-3-hydroxypyran-4-one-d3, 2-Methyl-3-hydroxypyrone-d3, 3-Hydroxy-2-methyl-1,4-pyrone-d3, Larixic Acid, Larixin-d3, NSC 2829-d3, NSC 404458-d3, Palatone-d3, Veltol-d3;
SMIL O=C1C=COC(C([2H])([2H])[2H])=C1O
Tekniske data
Kemisk navn eller materiale Maltol-d3
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C6H6O3/c1-4-6(8)5(7)2-3-9-4/h2-3,8H,1H3/i1D3
Procent renhed >95%