Få mere at vide
Malonic Acid-2-13C, TRC
Artikelnummer. 30475528
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475528 25 mg
30464549 50 mg
30470688 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475528 Leverandør Toronto Research Chemicals Leverandørnr. TRCM15800825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55514-11-9
Molekylær formel C213CH4O4
Molekylvægt (g/mol) 105.05
Synonym Propanedioic Acid-2-13C, 1,3-Propanedioic Acid-2-13C, Carboxyacetic Acid-2-13C, Dicarboxymethane-2-13C, Methanedicarboxylic Acid-2-13C, NSC 8124-2-13C, Malonic-2-13C Acid, [2-13C]Malonic Acid
SMIL OC([13C]([H])([H])C(O)=O)=O
Tekniske data
Kemisk navn eller materiale Malonic Acid-2-13C
Mængde 25 mg
InChI formel InChI=1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)/i1+1