missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid-2-13C, TRC

Product Code. 30464549
Change view
Click to view available options
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30464549 50 mg
30475528 25 mg
30470688 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464549 Supplier Toronto Research Chemicals Supplier No. TRCM15800850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55514-11-9
Molekylær formel C213CH4O4
Molekylvægt (g/mol) 105.05
Synonym Propanedioic Acid-2-13C, 1,3-Propanedioic Acid-2-13C, Carboxyacetic Acid-2-13C, Dicarboxymethane-2-13C, Methanedicarboxylic Acid-2-13C, NSC 8124-2-13C, Malonic-2-13C Acid, [2-13C]Malonic Acid
SMIL OC([13C]([H])([H])C(O)=O)=O

Specifications

Kemisk navn eller materiale Malonic Acid-2-13C
Mængde 50 mg
InChI formel InChI=1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)/i1+1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.