missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate

CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

225.00 DKK - 579.00 DKK

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11483257
SDS View Documents Promotion Details
Thermo Scientific Chemicals
A16186.14
25 g
225.00 DKK
25g
Please to purchase this item. Need a web account? Register with us today!
11493257
SDS View Documents
Thermo Scientific Chemicals
A16186.22
100 g
579.00 DKK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

Solubility
Soluble in water, alcohol, methanol, ethanol and ethylene glycol monomethyl ether.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong bases and strong acids.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Malachite Green oxalate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.