Få mere at vide
LY2795050, MedChemExpress
Artikelnummer. 16479307
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16479307 5 mg
16466886 100 mg
16466758 10 mg
16462837 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16479307 Leverandør Medchem Express Leverandørnr. HY15708
MedChemExpress LY2795050 is a novel selective κ-opioid Receptor (KOR) antagonist (IC50=0.72 nM) and has the potential as a PET tracer to image KOR in vivo.
LY2795050 is a novel selective κ-opioid Receptor (KOR) antagonist (IC50=0.72 nM) and has the potential as a PET tracer to image KOR in vivo.
Tekniske data
Kemisk navn eller materiale LY2795050
CAS 1346133-08-1
Farve White
Renhedsgrad noter Research
Molekylær formel C23H22ClN3O2
Mængde 5 mg
Opløselighedsinformation DMSO : 50 mg/mL (122.58 mM; Need ultrasonic)
SMIL O=C(N)C1=CC=C(OC2=CC=C(CN3[C@H](C4=CC=CN=C4)CCC3)C=C2)C(Cl)=C1
Molekylvægt (g/mol) 407.89
Formel vægt 407.89
Procent renhed 98.12%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation