Få mere at vide
LY2109761, MedChemExpress
Artikelnummer. 16487707
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16487707 5 mg
16455746 100 mg
16444978 10 mg
16411227 50 mg
16411036 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16487707 Leverandør Medchem Express Leverandørnr. HY12075
MedChemExpress LY2109761 is an orally active, selective TGF-β receptor type I/II inhibitor with Kis of 38 nM and 300 nM, respectively.
LY2109761 is an orally active, selective TGF-β receptor type I/II inhibitor with Kis of 38 nM and 300 nM, respectively.
Tekniske data
Kemisk navn eller materiale LY2109761
CAS 700874-71-1
Farve White
Renhedsgrad noter Research
Molekylær formel C26H27N5O2
Mængde 5 mg
Opløselighedsinformation DMSO : 25 mg/mL (56.62 mM; Need ultrasonic)
SMIL N12N=C(C(C3=C4C=CC(OCCN5CCOCC5)=CC4=NC=C3)=C1CCC2)C6=NC=CC=C6
Molekylvægt (g/mol) 441.52
Formel vægt 441.52
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease