missing translation for 'onlineSavingsMsg'
Learn More

Luminespib, MedChemExpress

Product Code. 16492836
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
10 mg
25 mg
100 mg
200 mg
500 mg
Unit Size:
10mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16492836 25 mg -
16484786 100 mg -
16453718 10 mg 10mg
16441546 500 mg -
16430956 200 mg -
16416457 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16492836 Supplier Medchem Express Supplier No. HY10215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Luminespib (VER-52296) is a potent HSP90 inhibitor with IC50s of 7.8 and 21 nM for HSP90α and HSP90β, respectively.

Luminespib (VER-52296) is a potent HSP90 inhibitor with IC50s of 7.8 and 21 nM for HSP90α and HSP90β, respectively.

Specifications

Kemisk navn eller materiale Luminespib
CAS 747412-49-3
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C26H31N3O5
Mængde 25 mg
Synonym VER-52296 AUY922 NVP-AUY922
Opløselighedsinformation DMSO : ≥ 62 mg/mL (133.18 mM)
SMIL O=C(C1=NOC(C2=CC(C(C)C)=C(O)C=C2O)=C1C3=CC=C(CN4CCOCC4)C=C3)NCC
Molekylvægt (g/mol) 465.54
Formel vægt 465.54
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.