Learn More
Lenz Laborglasinstrumente™ Low Shape Weighing Bottles
457.00 DKK - 1235.00 DKK
Specifications
Includes | Interchangeable NS stopper, inscription label |
---|---|
Product Line | Bottle |
Product Type | Weighing Bottle |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11758104
|
Lenz Laborglasinstrumente™
05565334 |
10 mL |
1180.00 DKK
Pack of 6 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
11768104
|
Lenz Laborglasinstrumente™
05565340 |
15 mL |
1235.00 DKK
Pack of 6 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
11826174
|
Lenz Laborglasinstrumente™
05565380 |
45 mL |
457.00 DKK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Specifications
Interchangeable NS stopper, inscription label | |
Bottle | |
Weighing Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Low Shape Weighing Bottles