Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
Brand:  Kautex™ 2000070536
 View more versions of this product
Product Code. 10380761
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Closures available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1500 mL
LDPE
No
Round
Wide Neck
1Each
No
Wide Neck Bottle
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kautex™ LDPE Wide Mouth Bottles without Closures > Capacity: 1500mL