Learn More
Lonza™ BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X)
Used as a buffer system for cell culture media. BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X) is manufactured in accordance with cGMP regulations and cell culture-tested to assure compatibility.
Brand: Lonza™ BE17-515F
Additional Details : Weight : 1.36080kg
Description
- Used to provide buffer system to maintain cell culture media in physiological pH range of 7.0 to 7.6
- Serves as irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for limited period of time
Specifications
15° to 30°C | |
No Calcium, No Magnesium | |
500 mL |
Dulbecco's Phosphate Buffered Saline | |
7 to 7.6 |