missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X)
Used as a buffer system for cell culture media. BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X) is manufactured in accordance with cGMP regulations and cell culture-tested to assure compatibility.
165.00 DKK - 308.00 DKK
Specifications
Content And Storage | 15° to 30°C |
---|---|
Product Type | Dulbecco's Phosphate Buffered Saline |
Without Additives | No Calcium, No Magnesium |
pH | 7 to 7.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11619980
|
Lonza™
BE17-515Q |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11609980
|
Lonza™
BE17-515F |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Used to provide buffer system to maintain cell culture media in physiological pH range of 7.0 to 7.6
- Serves as irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for limited period of time
Specifications
15° to 30°C | |
Dulbecco's Phosphate Buffered Saline | |
No Calcium, No Magnesium | |
7 to 7.6 |