Learn More
Lonza™ BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X)
Used as a buffer system for cell culture media.  BioWhittaker™ Dulbeccos Phosphate Buffered Saline (10X) is manufactured in accordance with cGMP regulations and cell culture-tested to assure compatibility.
Brand:  Lonza™ BE17-515F
Additional Details : Weight : 1.36080kg
 View more versions of this product
Product Code. 11609980
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Used to provide buffer system to maintain cell culture media in physiological pH range of 7.0 to 7.6
  • Serves as irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for limited period of time
Specifications
Specifications
15° to 30°C
No Calcium, No Magnesium
500 mL
Dulbecco's Phosphate Buffered Saline
7 to 7.6