Learn More
Lithium Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10605502
Lithium Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™
Product Code. p-7073268 Shop All Fisher Chemical Products
100 g, 100g
Click to view available options
Quantity:
100 g
250 g
5 kg
Unit Size:
100g
250g
5kg
Product Code. 10605502
Brand: Fisher Chemical L/2200/48
Request Bulk or Custom Format
Chemical Identifiers
CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]
Specifications
Boiling Point 1382°C
Melting Point 605°C
Color White
pH 7 to 8
Physical Form Solid
Quantity 100 g
Formula Weight 42.38g/mol
Packaging Plastic powder jar
Vapor Pressure 1.33 hPa 547
compliance-icons
Product Identifier
  • Lithium chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Product Title
Lithium Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™ >