Få mere at vide
Lercanidipine Hydrochloride, TRC
Artikelnummer. 30006977
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006977 50 mg
17973575 100 mg
17934515 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006977 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17900050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132866-11-6
Molekylær formel C36 H41 N3 O6 . Cl H
Molekylvægt (g/mol) 648.19
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3-[2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-methyl ester, hydrochloride (1:1), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester, monohydrochloride (9CI), Cardiovasc, Carmen, Corifeo, Lercadip, Lercan, Lercanidipine hydrochloride, Lercapin, Lercaton, Lerkamen, Lervasc, Lerzam, Lotensyl, R 75, Rec 15/2375, Renovia, Vasodip, Zandip, Zanicor
IUPAC navn 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMIL Cl.COC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C
Tekniske data
Kemisk navn eller materiale Lercanidipine Hydrochloride
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H
Formel vægt 647.2762
Procent renhed >95%