missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ LDPE Dropper Bottles

Use these transparent Vitlab™ LDPE Dropper Bottles – to store, save and dispense, using the extra long, fine dropping tip for accurate dispensing.

Brand:  Vitlab™ 94987

 View more versions of this product

Product Code. 10732361

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Narrow-mouth bottles
  • Made of LDPE
  • Transparent
  • With dropper insert
  • Screw cap made from LDPE
  • Extra long, fine dropping tip
Specifications

Specifications

250 mL
LDPE
12Pack
183 mm
58 mm
Transparent
12Pack
Narrow
Dropping Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vitlab™ LDPE Dropper Bottles > Capacity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.