Få mere at vide
Latanoprost Acid-d4(>85%), TRC
Artikelnummer. 30480242
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480242 10 mg
30479024 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480242 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17731210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1224443-47-3
Molekylær formel C23H30D4O5
Molekylvægt (g/mol) 394.54
Synonym (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoic-d4 Acid, 13,14-Dihydro-17-phenyl-18,19,20-trinor-PGF2α-d4, PhXA 85-d4, Latanoprost Related Compound E USP-d4
SMIL O[C@@H]1[C@H](C/C=C\C([2H])([2H])C([2H])([2H])CC(O)=O)[C@@H](CC[C@@H](O)CCC2=CC=CC=C2)[C@H](O)C1
Tekniske data
Kemisk navn eller materiale Latanoprost Acid-d4
Mængde 10 mg
InChI formel InChI=1S/C23H34O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,18-22,24-26H,2,7,10-16H2,(H,27,28)/b6-1-/t18-,19+,20+,21-,22+/m0/s1/i2D2,7D2