Få mere at vide
Larotrectinib Sulfate, TRC
Artikelnummer. 30475044
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475044 50 mg
30471685 250 mg
30474327 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475044 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17593350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1223405-08-0
Molekylær formel C21H22F2N6O2 . (H2O4S)
Molekylvægt (g/mol) 526.5
Synonym LOXO 101 sulfate, (3S)-N-[5-[(2R)-2-(2,5-Difluorophenyl)-1-pyrrolidinyl]pyrazolo[1,5-a]pyrimidin-3-yl]-3-hydroxy-1-pyrrolidinecarboxamide Sulfate
SMIL O[C@H]1CCN(C1)C(=O)Nc2cnn3ccc(nc23)N4CCC[C@@H]4c5cc(F)ccc5F.OS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Larotrectinib Sulfate
Mængde 50 mg
InChI formel InChI=1S/C21H22F2N6O2.H2O4S/c22-13-3-4-16(23)15(10-13)18-2-1-7-28(18)19-6-9-29-20(26-19)17(11-24-29)25-21(31)27-8-5-14(30)12-27;1-5(2,3)4/h3-4,6,9-11,14,18,30H,1-2,5,7-8,12H2,(H,25,31);(H2,1,2,3,4)/t14-,18+;/m0./s1