Få mere at vide
Invitrogen™ LanthaScreen™ Tb-pSer (PKC) Antibody Kit
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
Brand: Invitrogen™ PV3561
Beskrivelse
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
For use in combination with the Fluorescein-PKC substrate (Cat. No. PV3506) to evaluate inhibitors in medium to high throughput screening applications.
Tekniske data
| Tb (Terbium)/FITC | |
| Terbium-labeled anti-pSer (PKC Substrate) antibody in HEPES buffered saline. Store at -20°C. |
|
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| TR-FRET | |
| LanthaScreen |
| 1 mg | |
| 384-well Plate | |
| Human | |
| End Point | |
| Serine/Threonine Kinases | |
| AURKB, DYRK4, MARK4, MAP4K2, MAP4K4, MAP4K5, MINK1, MST4, NEK2, PHKG1, PKC, PRKX, ROCK2, RPS6KA1, RPS6KA2, STK24, STK25, SGK, SGK2, SGKL, TAOK2, TBK1 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
For Research Use Only. Not for use in diagnostic procedures.