Få mere at vide
Thermo Scientific™ LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
Brand: Thermo Scientific™ PV4751
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin
- This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines
- Use to develop robust, high-throughput biochemical or cell based assays for ubiquitination or deubiquitination
Tekniske data
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled anti-ubiquitin-FK2 antibody in HEPES buffered saline. Store at -20°C. | |
| 384-well Plate | |
| Microplate Reader | |
| Dry Ice | |
| LanthaScreen |
| 1 mg | |
| Liquid | |
| 340/490, 520 | |
| Human | |
| Hu |