Få mere at vide
Lamivudine (R)-Sulfoxide, TRC
Artikelnummer. 30475632
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475632 50 mg
30481637 10 mg
30475785 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475632 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17247550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 160552-54-5
Molekylær formel C8 H11 N3 O4 S
Molekylvægt (g/mol) 245.26
Synonym Lamivudine (R)-Sulfoxide, GI138870X
IUPAC navn 4-amino-1-[(2R,3R,5S)-2-(hydroxymethyl)-3-oxo-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMIL NC1=NC(=O)N(C=C1)[C@@H]2C[S@@](=O)[C@H](CO)O2
Tekniske data
Kemisk navn eller materiale 4-Amino-1-[(2R,3R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one S-Oxide
Mængde 50 mg
InChI formel InChI=1S/C8H11N3O4S/c9-5-1-2-11(8(13)10-5)6-4-16(14)7(3-12)15-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+,16+/m0/s1
Formel vægt 245.047