Learn More
Labconco™ Thimble Exhaust Connectors for Precise™ Glove Boxes
Accessories for use with Precise Glove Boxes
3395.00 DKK
Specifications
For Use With (Equipment) | Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box |
---|---|
Weight (English) | 5 lb. |
Weight (Metric) | 2.3 Kg |
Material | Epoxy-coated Steel |
Product Code | Brand | For Use With (Application) | Dimensions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Dimensions | Price | Quantity & Availability | ||||
16640463
|
Labconco™
5230200 |
Connecting the glove box to building exhaust or remote blower for outside ducting | 6 in. |
3395.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16650463
|
Labconco™
5241900 |
Connecting the glove box to the FilterMate™ portable exhauster | 5 in. |
3395.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Epoxy-coated steel for connecting the glove box to ductwork attached to building exhaust, remote blower or FilterMate Portable Exhauster. 5 in. diameter.Specifications
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box | |
5 lb. | |
2.3 Kg | |
Epoxy-coated Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Labconco™ Thimble Exhaust Connectors for Precise™ Glove Boxes