Learn More
MP Biomedicals LabCare Incubator Antimicrobial Spray
IVD-grade isothiazolone antimicrobial agents, highly efficient, eco-friendly and non-corrosive.
461.81 DKK - 4946.39 DKK
Specifications
Product Type | Disinfectant Spray |
---|---|
Color-coded | No |
Form | Liquid Spray |
Scent | Unscented |
Sterility | Non-sterile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30281213
|
MP Biomedicals
093051149 |
100 mL |
461.81 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30281208
|
MP Biomedicals
093051153 |
450 mL |
1283.23 DKK
450mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30281211
|
MP Biomedicals
093051153X4 |
1.8 L |
4946.39 DKK
1.80 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- MP Biomedicals LabCare Incubator Antimicrobial Spray is a broad-spectrum, efficient, stable, low-toxicity microbicide and preservative solution
- Specifically developed to inhibit and eliminate, Viruses, Bacteria, Fungi, Mycoplasma, and molds in incubators, biological safety cabinets, and ultra-clean benches
- This product effectively eliminates various microorganisms that affect cell culture, and the inhibitory effect on bacteria, fungi, spores, and conidia can last for 7-10 days
Specifications
Disinfectant Spray | |
Liquid Spray | |
Non-sterile |
No | |
Unscented |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
MP Biomedicals LabCare Incubator Antimicrobial Spray