Få mere at vide
L-Pyroglutamic Anhydride, TRC
Artikelnummer. 30008471
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008471 500 mg
30006516 250 mg
30003527 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008471 Leverandør Toronto Research Chemicals Leverandørnr. TRCP997485500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14842-41-2
Molekylær formel C10 H10 N2 O4
Molekylvægt (g/mol) 222.197
Synonym (5aS,10aS)-Dihydro-1H,5H-dipyrrolo[1,2-a:1',2'-d]pyrazine-3,5,8,10(2H,5aH,10aH)-tetrone, (-)-Dihydro-1H,5H-dipyrrolo[1,2-a:1',2'-d]pyrazine-3,5,8,10(2H,5aH,10aH)-tetrone, (5aS-cis)-Dihydro-1H,5H-dipyrrolo[1,2-a:1',2'-d]pyrazine-3,5,8,10(2H,5aH,10aH)-tetrone
IUPAC navn (5aS,10aS)-1,2,5a,6,7,10a-hexahydrodipyrrolo[1,3-c:1',3'-f]pyrazine-3,5,8,10-tetrone
SMIL O=C1CC[C@@H]2N1C(=O)[C@@H]3CCC(=O)N3C2=O
Tekniske data
Kemisk navn eller materiale L-Pyroglutamic Anhydride
InChI formel InChI=1S/C10H10N2O4/c13-7-3-1-5-9(15)12-6(2-4-8(12)14)10(16)11(5)7/h5-6H,1-4H2/t5-,6-/m0/s1
Mængde 500 mg
Formel vægt 222.0641