Få mere at vide
L-o-Tyrosine, TRC
Artikelnummer. 30453954
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453954 500 mg
30453080 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30453954 Leverandør Toronto Research Chemicals Leverandørnr. TRCT899990500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7423-92-9
Molekylær formel C9 H11 N O3
Molekylvægt (g/mol) 181.19
Synonym L-Phenylalanine, 2-hydroxy-, o-Tyrosine, L- (8CI), 2-Hydroxy-L-phenylalanine, (-)-o-Tyrosine, (S)-2-Amino-3-(2-hydroxyphenyl)propanoic acid, L-2-Hydroxyphenylalanine, L-3-(2-Hydroxyphenyl)alanine, L-o-Hydroxyphenylalanine, L-o-Tyrosine, o-L-Tyrosine, 2-Hydroxy-L-phenylalanine, (2S)-2-Amino-3-(2-hydroxyphenyl)propanoic acid, (S)-2-Amino-3-(2-hydroxyphenyl)propionic acid, L-α-Amino-β-(o-hydroxyphenyl)propionic acid
IUPAC navn (2S)-2-amino-3-(2-hydroxyphenyl)propanoic acid
SMIL N[C@@H](Cc1ccccc1O)C(=O)O
Tekniske data
Kemisk navn eller materiale (-)-o-Tyrosine
InChI formel InChI=1S/C9H11NO3/c10-7(9(12)13)5-6-3-1-2-4-8(6)11/h1-4,7,11H,5,10H2,(H,12,13)/t7-/m0/s1
Mængde 500 mg
Formel vægt 181.0739