Få mere at vide
L-Leucyl-D-leucine, TRC
Artikelnummer. 30482219
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482219 1 mg
30476181 2 mg
30470910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482219 Leverandør Toronto Research Chemicals Leverandørnr. TRCL3210201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17665-02-0
Molekylær formel C12H24N2O3
Molekylvægt (g/mol) 244.33
Synonym H-Leu-D-Leu-OH
IUPAC navn (2R)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-4-methylpentanoic acid
SMIL CC(C)C[C@H](N)C(=O)N[C@H](CC(C)C)C(=O)O
Tekniske data
Kemisk navn eller materiale L-Leucyl-D-leucine
InChI formel InChI=1S/C12H24N2O3/c1-7(2)5-9(13)11(15)14-10(12(16)17)6-8(3)4/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17)/t9-,10+/m0/s1
Mængde 1 mg
Formel vægt 244.1787