Få mere at vide
L-Hercynine, TRC
Artikelnummer. 30136942
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136942 250 mg
30138518 500 mg
30012621 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136942 Leverandør Toronto Research Chemicals Leverandørnr. TRCH288900250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 534-30-5
Molekylær formel C9 H15 N3 O2
Molekylvægt (g/mol) 197.23
Synonym 1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (αS)-, 1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (S)-, Ammonium, (1-carboxy-2-imidazol-4-ylethyl)trimethyl-, hydroxide, inner salt, L- (8CI), (2S)-3-(1H-Imidazol-4-yl)-2-(trimethylazaniumyl)propanoate, (2S)-3-(1H-Imidazol-5-yl)-2-(trimethylazaniumyl)propanoate, Histidine-betaine, N-α-trimethyl histidine, α-N,N,N-Trimethylhistidine, L-Hercynine
IUPAC navn (2S)-3-(1H-imidazol-5-yl)-2-(trimethylazaniumyl)propanoate
SMIL C[N+](C)(C)[C@@H](Cc1cnc[nH]1)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale L-Hercynine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H15N3O2/c1-12(2,3)8(9(13)14)4-7-5-10-6-11-7/h5-6,8H,4H2,1-3H3,(H-,10,11,13,14)/t8-/m0/s1
Mængde 250 mg
Formel vægt 197.1164
Procent renhed >95%