CAS RN 534-30-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
L-Hercynine, TRC
CAS: 534-30-5 Molekylær formel: C9 H15 N3 O2 Molekylvægt (g/mol): 197.23 Synonym: 1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (αS)-,1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (S)-,Ammonium, (1-carboxy-2-imidazol-4-ylethyl)trimethyl-, hydroxide, inner salt, L- (8CI),(2S)-3-(1H-Imidazol-4-yl)-2-(trimethylazaniumyl)propanoate,(2S)-3-(1H-Imidazol-5-yl)-2-(trimethylazaniumyl)propanoate,Histidine-betaine,N-α-trimethyl histidine,α-N,N,N-Trimethylhistidine,L-Hercynine IUPAC navn: (2S)-3-(1H-imidazol-5-yl)-2-(trimethylazaniumyl)propanoate SMIL: C[N+](C)(C)[C@@H](Cc1cnc[nH]1)C(=O)[O-]