missing translation for 'onlineSavingsMsg'
Få mere at vide

L-Felinine, TRC

Artikelnummer. 30240945
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240945 5 mg
30240978 50 mg
30240848 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30240945 Leverandør Toronto Research Chemicals Leverandørnr. TRCF2312505MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 471-09-0
Molekylær formel C8 H17 N O3 S
Molekylvægt (g/mol) 207.29
IUPAC navn (2R)-2-amino-3-(4-hydroxy-2-methylbutan-2-yl)sulfanylpropanoic acid
SMIL CC(C)(CCO)SC[C@H](N)C(=O)O

Tekniske data

Kemisk navn eller materiale L-Felinine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H17NO3S/c1-8(2,3-4-10)13-5-6(9)7(11)12/h6,10H,3-5,9H2,1-2H3,(H,11,12)/t6-/m0/s1
Mængde 5 mg
Formel vægt 207.0929
Procent renhed >95%
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.