Få mere at vide
L-Cysteinylglycine, TRC
Artikelnummer. 30137496
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137496 100 mg
30134646 25 mg
30008879 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137496 Leverandør Toronto Research Chemicals Leverandørnr. TRCC994770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19246-18-5
Synonym N-L-Cysteinylglycine∣144: PN: US20130123467 SEQID: 173 Claimed Protein∣145: PN: WO2012013136 SEQID: 39 Unclaimed Protein∣27: PN: WO2011133608 TABLE: 5 Claimed Sequence∣52: PN: EP2161028 PAGE: 10 Claimed Protein∣6: PN: WO2011003622 SEQID: 6 Claimed Protein∣Cysteinylglycine∣L-Cysteinylglycine;
Tekniske data
Renhed >95% (HPLC)
Format Neat
Molekylær formel C5H10N2O3S
MDL nummer MFCD00055567
Molekylvægt (g/mol) 178.21
InChI nøgle ZUKPVRWZDMRIEO-VKHMYHEASA-N
SMIL N[C@@H](CS)C(=O)NCC(O)=O
IUPAC navn 2-[(2R)-2-amino-3-sulfanylpropanamido]acetic acid