CAS RN 19246-18-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
L-Cysteinylglycine, TRC
CAS: 19246-18-5 Synonym: N-L-Cysteinylglycine∣144: PN: US20130123467 SEQID: 173 Claimed Protein∣145: PN: WO2012013136 SEQID: 39 Unclaimed Protein∣27: PN: WO2011133608 TABLE: 5 Claimed Sequence∣52: PN: EP2161028 PAGE: 10 Claimed Protein∣6: PN: WO2011003622 SEQID: 6 Claimed Protein∣Cysteinylglycine∣L-Cysteinylglycine;