Få mere at vide
L-Argininamide-D-phenylalanyl-L-2-piperidinecarbonyl-N-(4-nitrophenyl) Acetic Acid (D-PHE-PIP-ARG-PNA), TRC
Artikelnummer. 30423109
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30423109 5 mg -
30421520 1 mg -
30421326 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423109 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3363855MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 386253-60-7
Molekylær formel C27H36N8O5 • x(C2H4O2)
Molekylvægt (g/mol) 672.7
Synonym L-Argininamide, D-phenylalanyl-(2S)-2-piperidinecarbonyl-N-(4-nitrophenyl)-triacetate, H-D-Phe-Homopro-Arg-pNA.2 Acetate
IUPAC navn (S)-1-(D-phenylalanyl)-N-((S)-5-guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)piperidine-2-carboxamide acetate
SMIL O=C([C@H](CCCNC(N)=N)NC([C@H]1N(C([C@@H](CC2=CC=CC=C2)N)=O)CCCC1)=O)NC3=CC=C([N+]([O-])=O)C=C3.OC(C)=O
Tekniske data
Kemisk navn eller materiale L-Argininamide-D-phenylalanyl-L-2-piperidinecarbonyl-N-(4-nitrophenyl) Acetic Acid (D-PHE-PIP-ARG-PNA)
Mængde 5 mg
InChI formel InChI=1S/C27H36N8O5.C2H4O2/c28-21(17-18-7-2-1-3-8-18)26(38)34-16-5-4-10-23(34)25(37)33-22(9-6-15-31-27(29)30)24(36)32-19-11-13-20(14-12-19)35(39)40;1-2(3)4/h1-3,7-8,11-14,21-23H,4-6,9-10,15-17,28H2,(H,32,36)(H,33,37)(H4,29,30,31);1H3,(H,3,4)/t21-,22+,23+;/m1./s1