Få mere at vide
Tocris Bioscience™ L 006235
GREENER_CHOICE
Artikelnummer. 18055082
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18055082 10 mg
18084642 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18055082 Leverandør Tocris Bioscience™ Leverandørnr. 3066/10
Potent cathepsin K inhibitor
Potent, reversible cathepsin K inhibitor (IC50 = 0.25 nM) that displays > 4000-fold selectivity over cathepsins B, L and S. Displays reduced selectivity in cell-based assays possibly due to lysosomal accumulation. Reduces collagen breakdown and promotes bone deposition in vivo. Orally active and has intrinsic fluorescence.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 294623-49-7
Molekylær formel C24H30N6O2S
Tekniske data
Kemisk navn eller materiale L 006235
Mængde 10 mg
Formel vægt 466.6
Renhed >98%
Molekylvægt (g/mol) 466.604