Få mere at vide
KRN2 bromide, MedChemExpress
Artikelnummer. 16477287
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16477287 5 mg
16463136 25 mg
16445426 100 mg
16430877 50 mg
16414578 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16477287 Leverandør Medchem Express Leverandørnr. HY112125A
MedChemExpress KRN2 (bromide) is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM.
KRN2 (bromide) is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM.
Tekniske data
Kemisk navn eller materiale KRN2 bromide
CAS 1390654-28-0
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C27H23BrFNO4
Mængde 5 mg
Opløselighedsinformation DMSO : 18.33 mg/mL (34.96 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL COC1=C(OC)C2=C[N+]3=C(C(C(CC3)=C4)=CC5=C4OCO5)C(CC6=CC=CC=C6F)=C2C=C1.[Br-]
Molekylvægt (g/mol) 524.38
Formel vægt 524.38
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation