missing translation for 'onlineSavingsMsg'
Få mere at vide

KML29, MedChemExpress

Artikelnummer. 16487668
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16487668 10 mg
16483677 50 mg
16430138 5 mg
16407576 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16487668 Leverandør Medchem Express Leverandørnr. HY18977

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress KML29 is an extremely selective, orally active and irreversible MAGL inhibitor, with IC50 values of 15 nM, 43 nM and 5.9 nM for mouse, rat and human MAGL, respectively. KML29 exhibits minimal cross-reactivity toward other central and peripheral serine hydrolases, including no detectable activity against FAAH.

KML29 is an extremely selective, orally active and irreversible MAGL inhibitor, with IC50 values of 15 nM, 43 nM and 5.9 nM for mouse, rat and human MAGL, respectively. KML29 exhibits minimal cross-reactivity toward other central and peripheral serine hydrolases, including no detectable activity against FAAH.

Tekniske data

Kemisk navn eller materiale KML29
CAS 1380424-42-9
Farve White
Renhedsgrad noter Research
Molekylær formel C24H21F6NO7
Mængde 10 mg
Opløselighedsinformation DMSO : 50 mg/mL (91.01 mM; Need ultrasonic)
SMIL O=C(N1CCC(C(C2=CC=C(OCO3)C3=C2)(C4=CC=C(OCO5)C5=C4)O)CC1)OC(C(F)(F)F)C(F)(F)F
Molekylvægt (g/mol) 549.42
Formel vægt 549.42
Procent renhed 98.77%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.