Få mere at vide
Kimberly-Clark Professional™ Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
Ring for pris
Advarsel:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Produkter 1
Produktkode Brand DimensionsLxW PackagingQuantity Pris Antal & tilgængelighed  
Produktkode Brand DimensionsLxW PackagingQuantity Pris Antal & tilgængelighed  
15142193
Dokumentvisning
Kimberly-Clark Professional™
34155
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ikke relevant Se alternative produkter
Spar op til 
»
Beskrivelse
Beskrivelse
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
  • Single ply, light-duty wiper that can handle a variety of delicate tasks
  • Extra low-lint wipers made from 100% virgin wood fibers
  • Soft, nonabrasive cellulose-fiber wipers won't scratch most delicate surfaces
  • Easily wipe up liquid and dust
  • Absorb many times their own weight
  • Handy one-at-a-time dispensing packs with special plastic guard to reduce airborne lint, keep contaminants out of box and reduce electrostatic discharge
Videoer
SDS
Dokumenter
Dokumenter
Produktcertifikater