Få mere at vide
Kimberly-Clark Professional™ Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
Brand: Kimberly-Clark Professional™ 34155
Advarsel:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Beskrivelse
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
- Single ply, light-duty wiper that can handle a variety of delicate tasks
- Extra low-lint wipers made from 100% virgin wood fibers
- Soft, nonabrasive cellulose-fiber wipers won't scratch most delicate surfaces
- Easily wipe up liquid and dust
- Absorb many times their own weight
- Handy one-at-a-time dispensing packs with special plastic guard to reduce airborne lint, keep contaminants out of box and reduce electrostatic discharge