Learn More
Ketorolac tromethamine, Thermo Scientific™
605.00 DKK - 1770.00 DKK
Chemical Identifiers
CAS | 74103-07-4 |
---|---|
Molecular Formula | C19H24N2O6 |
Molecular Weight (g/mol) | 376.41 |
MDL Number | 00887595 |
InChI Key | BWHLPLXXIDYSNW-UHFFFAOYNA-N |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid |
SMILES | NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17565575
|
Thermo Scientific Acros
469170010 |
1 g |
605.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17575575
|
Thermo Scientific Acros
469170050 |
5 g |
1770.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
74103-07-4 | |
376.41 | |
BWHLPLXXIDYSNW-UHFFFAOYNA-N | |
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 |
C19H24N2O6 | |
00887595 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid |
Specifications
Glass Bottle | |
Available | |
C19H24N2O6 | |
White to Light Yellow | |
13-5324 | |
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1 | |
376.41 |
74103-07-4 | |
Powder | |
00887595 | |
160°C to 165°C | |
BWHLPLXXIDYSNW-UHFFFAOYNA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid | |
Ketorolac tromethamine |
Safety and Handling
Skull and crossbones
missing translation for 'rtecsNumber' : UY7759900
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ketorolac tromethamine, Thermo Scientific™