missing translation for 'onlineSavingsMsg'
Få mere at vide

Ketoprofen, Thermo Scientific™

A non-steroidal anti-inflammatory drug (NSAID); also inhibits cyclooxygenases-1 and -2

731.35 DKK - 2313.47 DKK

Kemiske identifikatorer

CAS 22071-15-4
Molekylær formel C16H14O3
Molekylvægt (g/mol) 254.285
MDL nummer MFCD00055790
InChI nøgle DKYWVDODHFEZIM-UHFFFAOYSA-N
Synonym ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
PubChem CID 3825
ChEBI CHEBI:6128
IUPAC navn 2-(3-benzoylphenyl)propanoic acid
SMIL CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O
Se flere specifikationer

Produkter 2
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15495019
Dokumentvisning
Thermo Scientific Alfa Aesar
J62702.06
5g
731.35 DKK
5g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15405029
Dokumentvisning
Thermo Scientific Alfa Aesar
J62702.14
25g
2313.47 DKK
25g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a anti-inflammatory; analgesic and an antipyretic drug cause the hypothalamus to override an interleukin-induced increase in temperature. They act by blocking the synthesis of prostaglandins by inhibiting cyclooxygenase, which converts arachidonic acid to cyclic endoperoxides, precursors of prostaglandins.

Solubility
Slightly soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Tekniske data

Kemiske identifikatorer

22071-15-4
254.285
DKYWVDODHFEZIM-UHFFFAOYSA-N
3825
2-(3-benzoylphenyl)propanoic acid
C16H14O3
MFCD00055790
ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
CHEBI:6128
CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O

Tekniske data

Ketoprofen
C16H14O3
UN2811
ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat
DKYWVDODHFEZIM-UHFFFAOYSA-N
2-(3-benzoylphenyl)propanoic acid
3825
254.29
22071-15-4
MFCD00055790
14,5305
Slightly soluble in chloroform and methanol.
CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O
254.285
CHEBI:6128
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 244-759-8

missing translation for 'rtecsNumber' : UE7570000

missing translation for 'tsca' : Yes

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.