missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
KAT4/TBP Associated Factor 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1455.00 DKK - 3580.00 DKK
Tekniske data
| Antigen | KAT4/TBP Associated Factor 1 |
|---|---|
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Form | Purified |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18306206
|
Novus Biologicals
NBP3-03499-20UL |
20 μL |
1455.00 DKK
20µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18310754
|
Novus Biologicals
NBP3-03499-100UL |
100 μL |
3580.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
KAT4/TBP Associated Factor 1 Polyclonal specifically detects KAT4/TBP Associated Factor 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| KAT4/TBP Associated Factor 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1893 of human KAT4/TBP Associated Factor 1 (NP_004597.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 50% glycerol, pH7.3. | |
| 6872 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel