missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
KAT4/TBP Associated Factor 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03499-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KAT4/TBP Associated Factor 1 Polyclonal specifically detects KAT4/TBP Associated Factor 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| KAT4/TBP Associated Factor 1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1893 of human KAT4/TBP Associated Factor 1 (NP_004597.2). | |
| 20 μL | |
| Protein Kinase | |
| 6872 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion