missing translation for 'onlineSavingsMsg'
Få mere at vide

JMV 2959, MedChemExpress

Artikelnummer. 16484096
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16484096 25 mg
16472638 5 mg
16460439 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16484096 Leverandør Medchem Express Leverandørnr. HYU00433

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress JMV 2959 is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32 nM.

JMV 2959 is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32 nM.

Tekniske data

Kemisk navn eller materiale JMV 2959
CAS 925238-89-7
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C30H32N6O2
Mængde 25 mg
Opløselighedsinformation DMSO : 25 mg/mL (49.15 mM; ultrasonic and warming and heat to 60°C)
SMIL O=C(N[C@@H](C1=NN=C(CCC2=CC=CC=C2)N1CC3=CC=C(OC)C=C3)CC4=CNC5=C4C=CC=C5)CN
Molekylvægt (g/mol) 508.61
Formel vægt 508.61
Procent renhed 99.91%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.